Tisk…

1-Hexylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Hexylmagnesium chloride

M-Clarity™ quality level = MQ100

641626-100ML 641626-800ML
SIAL641626-100MLEA 4610 CZK
SIAL641626-100ML SIAL641626-800ML
1-Hexylmagnesium chloride 2 mol/l in THF, Sigma-Aldrich®
1-Hexylmagnesium chloride
Vzorec: C₆H₁₃ClMg
MDL Number: MFCD06798087
CAS číslo: 44767-62-6
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR