Tisk…

3-Methylbenzo[b]thiophene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

638587-1G 638587-5G
SIAL638587-1GEA 1920 CZK
SIAL638587-1G SIAL638587-5G
3-Methylbenzo[b]thiophene, Sigma-Aldrich®
3-Methylbenzo[b]thiophene
Vzorec: C₉H₈S
MDL Number: MFCD00040243
CAS číslo: 1455-18-1
EINECS: 215-934-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR