Tisk…

(S)-(-)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

634794-1G 634794-5G
SIAL634794-1GEA 4200 CZK
SIAL634794-1G SIAL634794-5G
(S)-(-)-1-Boc-3-aminopyrrolidine, Sigma-Aldrich®
(S)-(-)-1-Boc-3-aminopyrrolidine
Vzorec: C₉H₁₈N₂O₂
MDL Number: MFCD03419271
CAS číslo: 147081-44-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR