Tisk…

(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

631310-1G 631310-5G
SIAL631310-1GEA 2890 CZK
SIAL631310-1G SIAL631310-5G
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid, Sigma-Aldrich®
(3,5-Dimethoxy-4-hydroxyphenyl)acetic acid
Vzorec: C₁₀H₁₂O₅
MDL Number: MFCD00016834
CAS číslo: 4385-56-2
EINECS: 224-494-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR