Tisk…

Indole-6-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1H-Indole-6-carboxylic acid

576662-5G
SIAL576662-5GEA 4950 CZK
SIAL576662-5G
Indole-6-carboxylic acid, Sigma-Aldrich®
Indole-6-carboxylic acid
Vzorec: C₉H₇NO₂
MW: 161,16 g/mol
Bod tání: 254…257 °C
MDL Number: MFCD00210441
CAS číslo: 1670-82-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR