Tisk…

3-Pyridineboronic acid pinacol ester, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

576565-1G 576565-5G
SIAL576565-1GEA 2650 CZK
SIAL576565-1G SIAL576565-5G
3-Pyridineboronic acid pinacol ester, Sigma-Aldrich®
3-Pyridineboronic acid pinacol ester
Vzorec: C₁₁H₁₆BNO₂
MDL Number: MFCD03084758
CAS číslo: 329214-79-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR