Tisk…

4,4'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

569593-1G 569593-5G
SIAL569593-1GEA 861 CZK
SIAL569593-1G SIAL569593-5G
4,4'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
4,4'-Dimethyl-2,2'-bipyridyl
Vzorec: C₁₂H₁₂N₂
MDL Number: MFCD00006441
CAS číslo: 1134-35-6
EINECS: 214-483-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR