Tisk…

4-(Bromomethyl)benzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-(Bromomethyl)benzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

557153-5G
SIAL557153-5GEA 4070 CZK
SIAL557153-5G
4-(Bromomethyl)benzenesulphonyl chloride, Sigma-Aldrich®
4-(Bromomethyl)benzenesulphonyl chloride
Vzorec: C₇H₆BrClO₂S
MW: 269.55
Bod tání: 74…76 °C
MDL Number: MFCD00156129
CAS číslo: 66176-39-4
UN: 3261
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR