Tisk…

1-(Heptafluorobutyryl)imidazole, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

556645-1G 556645-5G
SIAL556645-1GEA 2470 CZK
SIAL556645-1G SIAL556645-5G
1-(Heptafluorobutyryl)imidazole, Sigma-Aldrich®
1-(Heptafluorobutyryl)imidazole
Vzorec: C₇H₃F₇N₂O
MDL Number: MFCD00014503
CAS číslo: 32477-35-3
EINECS: 251-063-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR