Tisk…

Oxid hlinitý, nanopowder <50 nm (TEM), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

M-Clarity™ quality level = MQ200

544833-10G 544833-50G
SIAL544833-10GEA 1350 CZK
SIAL544833-10G SIAL544833-50G
Oxid hlinitý, nanopowder <50 nm (TEM), Sigma-Aldrich®
Oxid hlinitý
Vzorec: Al₂O₃
MW: 101.96
Bod varu: 2980
Bod tání: 2045
Hustota: 3.94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS číslo: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR