Tisk…

(±)-4-Chlorostyrene oxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-(4-Chlorophenyl)oxirane

M-Clarity™ quality level = MQ100

534757-5G
SIAL534757-5GEA 2510 CZK
SIAL534757-5G
(±)-4-Chlorostyrene oxide, Sigma-Aldrich®
(±)-4-Chlorostyrene oxide
Vzorec: C₈H₇ClO
MW: 154.6
MDL Number: MFCD00956179
CAS číslo: 2788-86-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR