Tisk…

Palladium(II) acetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Palladium diacetate, Palladium (II) acetate

M-Clarity™ quality level = MQ200

520764-1G 520764-25G 520764-5G
SIAL520764-1GEA 3820 CZK
SIAL520764-1G SIAL520764-25G SIAL520764-5G
Palladium(II) acetate, Sigma-Aldrich®
Palladium(II) acetate
Vzorec: C₄H₆O₄Pd
Bod tání: 205 °C
MDL Number: MFCD00012453
CAS číslo: 3375-31-3
EINECS: 222-164-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR