Tisk…

Hemin chloride (from porcine) ≥96.0% (by HPLC), BioXtra, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

51280-1G 51280-25G 51280-5G
SIAL51280-1GEA 2350 CZK
SIAL51280-1G SIAL51280-25G SIAL51280-5G
Hemin chloride (from porcine) ≥96.0% (by HPLC), BioXtra, Sigma-Aldrich®
Hemin chloride
Vzorec: C₃₄H₃₂ClFeN₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00010726
CAS číslo: 16009-13-5
EINECS: 240-140-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR