Tisk…

Glutaraldehyd 50% in aqueous solution, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial

M-Clarity™ quality level = MQ100

49629-1L 49629-2.5L 49629-250ML
SIAL49629-1LEA 3780 CZK
SIAL49629-1L SIAL49629-2.5L SIAL49629-250ML
Glutaraldehyd 50% in aqueous solution, Sigma-Aldrich®
Glutaraldehyd
Vzorec: C₅H₈O₂
Hustota: 1,13 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS číslo: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR