Tisk…

Chloroform-D1 for NMR spectroscopy, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Trichloromethane-D1, Chloroform-[²H₁], Chloroform-D

M-Clarity™ quality level = MQ200

494275-10G 494275-50G
SIAL494275-10GEA 2120 CZK
SIAL494275-10G SIAL494275-50G
Chloroform-D1 for NMR spectroscopy, Sigma-Aldrich®
Chloroform-D1
Vzorec: CDCl₃
Bod varu: 60,8 °C (1013 hPa)
Hustota: 1,5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS číslo: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR