Tisk…

2-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

491624-10G
SIAL491624-10GEA 3250 CZK
SIAL491624-10G
2-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
2-(Trifluoromethyl)benzenesulphonyl chloride
Vzorec: C₇H₄ClF₃O₂S
Bod varu: 123…126 °C (1013 hPa)
MDL Number: MFCD00051696
CAS číslo: 776-04-5
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR