Tisk…

Devardova slitina, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Copper aluminium zinc alloy (50:45:5 w%), Devarda's metal

M-Clarity™ quality level = MQ100

483907-100G
SIAL483907-100GEA 3280 CZK
SIAL483907-100G
Devardova slitina, Sigma-Aldrich®
Devardova slitina
Bod varu: 906 °C (1013 hPa)
Bod tání: 490…560 °C
Hustota: 5,79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198176
CAS číslo: 8049-11-4
UN: 3089
ADR: 4.1,II
Merck Index: 13,02958

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR