Tisk…

Benzoin methyl ether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,2-Diphenyl-2-methoxyethan-1-one, 2-Methoxy-1,2-diphenylethanone

M-Clarity™ quality level = MQ200

479314-25G
SIAL479314-25GEA 4830 CZK
SIAL479314-25G
Benzoin methyl ether, Sigma-Aldrich®
Benzoin methyl ether
Vzorec: C₁₅H₁₄O₂
MW: 226.27
MDL Number: MFCD00008492
CAS číslo: 3524-62-7
EINECS: 222-538-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR