Tisk…

tert-Butylchlorodimethylsilane 50% in toluene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

473464-100G 473464-50G
SIAL473464-100GEA 4990 CZK
SIAL473464-100G SIAL473464-50G
tert-Butylchlorodimethylsilane 50% in toluene, Sigma-Aldrich®
tert-Butylchlorodimethylsilane
Vzorec: C₆H₁₅ClSi
MDL Number: MFCD00000501
CAS číslo: 18162-48-6
EINECS: 242-042-4
UN: 2924
ADR: 3,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR