Tisk…

4-Fluoro-7-nitrobenzofurazan, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole, NBD-F

M-Clarity™ quality level = MQ200

47140-10MG 47140-50MG
SIAL47140-10MGEA 9630 CZK
SIAL47140-10MG SIAL47140-50MG
4-Fluoro-7-nitrobenzofurazan, Sigma-Aldrich®
4-Fluoro-7-nitrobenzofurazan
Vzorec: C₆H₂FN₃O₃
MW: 183.1
MDL Number: MFCD00010196
CAS číslo: 29270-56-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR