Tisk…

Lithium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

451622-25G 451622-5G
SIAL451622-25GEA 19030 CZK
SIAL451622-25G SIAL451622-5G
Lithium tetrafluoroborate, Sigma-Aldrich®
Lithium tetrafluoroborate
Vzorec: BF₄Li
Hustota: 0,852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS číslo: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR