Tisk…

Iron(II) chloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí

Synonyms: Iron dichloride, Ferrous chloride

M-Clarity™ quality level = MQ100

450944-10G 450944-50G
SIAL450944-10GEA 5050 CZK
SIAL450944-10G SIAL450944-50G
Iron(II) chloride, Sigma-Aldrich®
Iron(II) chloride
Vzorec: Cl₂Fe
Bod varu: 2023 °C (1013 hPa)
Bod tání: 674 °C
Hustota: 2,7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS číslo: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8,III
Merck Index: 12,04091

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR