Tisk…

Gadolinium trichloride hydrate, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Gadolinium(III) chloride hydrate

450855-10G 450855-50G
SIAL450855-10GEA 3670 CZK
SIAL450855-10G SIAL450855-50G
Gadolinium trichloride hydrate, Sigma-Aldrich®
Gadolinium trichloride hydrate
Vzorec: Cl₃Gd·H₂O
MDL Number: MFCD00149732
CAS číslo: 19423-81-5
EINECS: 233-386-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR