Tisk…

Pluronic® L-64, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

435449-1L 435449-250ML
SIAL435449-1LEA 4640 CZK
SIAL435449-1L SIAL435449-250ML
Pluronic® L-64, Sigma-Aldrich®
Poloxamer 184 (Pluronic® L-64, Synperonic® L64)

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Vzorec: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
Storage Temperature: Ambient
MDL Number: MFCD00082049
CAS číslo: 9003-11-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR