Tisk…

Bis[N,N-bis(trimethylsilyl)amide]tin(II), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

409618-25ML
SIAL409618-25MLEA 8420 CZK
SIAL409618-25ML
Bis[N,N-bis(trimethylsilyl)amide]tin(II), Sigma-Aldrich®
Bis[N,N-bis(trimethylsilyl)amide]tin(II)
Vzorec: C₁₂H₃₆N₂Si₄Sn
MDL Number: MFCD00191794
CAS číslo: 59863-13-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR