Tisk…

Dipotassium tellurium tetraoxide hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

400580-25G
SIAL400580-25GEA 3650 CZK
SIAL400580-25G
Dipotassium tellurium tetraoxide hydrate, Sigma-Aldrich®
Dipotassium tellurium tetraoxide hydrate
Vzorec: K₂O₄Te·H₂O
CAS číslo: 314041-10-6
EINECS: 239-626-6
UN: 3284
ADR: 6.1,II
Merck Index: 12,07851

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR