Tisk…

4-(Methylthio)phenylacetic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

396591-1G
SIAL396591-1GEA 4080 CZK
SIAL396591-1G
4-(Methylthio)phenylacetic acid, Sigma-Aldrich®
4-(Methylthio)phenylacetic acid
Vzorec: C₉H₁₀O₂S
MW: 182.24
MDL Number: MFCD00192325
CAS číslo: 16188-55-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR