Tisk…

ethylamin 2 mol/l in methanol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

395064-100ML 395064-800ML
SIAL395064-100MLEA 1690 CZK
SIAL395064-100ML SIAL395064-800ML
ethylamin 2 mol/l in methanol, Sigma-Aldrich®
ethylamin
Vzorec: C₂H₇N
MDL Number: MFCD00008160
CAS číslo: 75-04-7
EINECS: 200-834-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR