Tisk…

3-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-(Trifluoromethyl)benzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

385417-5G
SIAL385417-5GEA 1760 CZK
SIAL385417-5G
3-(Trifluoromethyl)benzenesulphonyl chloride, Sigma-Aldrich®
3-(Trifluoromethyl)benzenesulphonyl chloride
Vzorec: C₇H₄ClF₃O₂S
MW: 244.62
Bod varu: 75…76 °C (0.5 mmHg)
Hustota: 1.52
MDL Number: MFCD00014724
CAS číslo: 777-44-6
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR