Tisk…

Hydrogenfosforečnan diamonný, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate

M-Clarity™ quality level = MQ100

379980-100G 379980-25G
SIAL379980-100GEA 12400 CZK
SIAL379980-100G SIAL379980-25G
Hydrogenfosforečnan diamonný, Sigma-Aldrich®
Hydrogenfosforečnan diamonný
Vzorec: (NH₄)₂HPO₄
Bod tání: 185 °C
Hustota: 1,619 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010891
CAS číslo: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Index: 13,00546

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR