Tisk…

Titanium(IV) isopropoxide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide

M-Clarity™ quality level = MQ200

377996-100ML 377996-25ML 377996-5ML
SIAL377996-100MLEA 20250 CZK
SIAL377996-100ML SIAL377996-25ML SIAL377996-5ML
Titanium(IV) isopropoxide, Sigma-Aldrich®
Titanium(IV) isopropoxide
Vzorec: C₁₂H₂₈O₄Ti
MW: 284.23
Bod varu: 240
Bod tání: 18…20 °C
Hustota: 0.9711
Bod vzplanutí: 51
MDL Number: MFCD00008871
CAS číslo: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3 III
Merck Index: 12,09614

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR