Tisk…

1-Fluoropyridiniumtetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: n-Fluoropyridinium tetrafluoroborate

MQ100

377260-1G
SIAL377260-1GEA 0 CZK
SIAL377260-1G
1-Fluoropyridiniumtetrafluoroborate, Sigma-Aldrich®
1-Fluoropyridiniumtetrafluoroborate
Vzorec: C₅H₅BF₅N
MW: 184.9
MDL Number: MFCD00153176
CAS číslo: 107264-09-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR