Tisk…

Boron trifluoride dihydrate, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

359963-100ML 359963-500ML
SIAL359963-100MLEA 1960 CZK
SIAL359963-100ML SIAL359963-500ML
Boron trifluoride dihydrate, Sigma-Aldrich®
Boron trifluoride dihydrate
Vzorec: BF₃·2H₂O
Bod varu: 58,5…60 °C (1 mmHg)
Hustota: 1,636 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149901
CAS číslo: 13319-75-0
EINECS: 231-569-5
UN: 2851
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR