Tisk…

Perfluoroheptanoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

342041-25G 342041-5G
SIAL342041-25GEA 4670 CZK
SIAL342041-25G SIAL342041-5G
Perfluoroheptanoic acid, Sigma-Aldrich®
Perfluoroheptanoic acid
Vzorec: C₇HF₁₃O₂
MDL Number: MFCD00039604
CAS číslo: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR