Tisk…

4-Aminoantipyrin, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone, 4-Aminophenazone

M-Clarity™ quality level = MQ200

33528-100G-R 33528-25G-R
SIAL33528-100G-REA 5100 CZK
SIAL33528-100G-R SIAL33528-25G-R
4-Aminoantipyrin, Sigma-Aldrich®
4-Aminoantipyrin
Vzorec: C₁₁H₁₃N₃O
MW: 203.24
Bod tání: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS číslo: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR