Tisk…

4-Sulphobenzoic acid monopotassium salt, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Sulphobenzoic acid potassium salt

M-Clarity™ quality level = MQ200

310638-50G
SIAL310638-50GEA 1750 CZK
SIAL310638-50G
4-Sulphobenzoic acid monopotassium salt, Sigma-Aldrich®
4-Sulphobenzoic acid monopotassium salt
Vzorec: C₇H₅KO₅S
MDL Number: MFCD00007509
CAS číslo: 5399-63-3
EINECS: 226-435-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR