Tisk…

Rhodium(III) chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

307866-2.5G 307866-500MG
SIAL307866-2.5GEA 67500 CZK
SIAL307866-2.5G SIAL307866-500MG
Rhodium(III) chloride, Sigma-Aldrich®
Rhodium(III) chloride
Vzorec: Cl₃Rh
Hustota: 5,38 g/cm³ (20 °C)
MDL Number: MFCD00011204
CAS číslo: 10049-07-7
EINECS: 233-165-4
Merck Index: 12,08355

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR