Tisk…

Propylphosphonic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

305685-1G
SIAL305685-1GEA 2510 CZK
SIAL305685-1G
Propylphosphonic acid, Sigma-Aldrich®
Propylphosphonic acid
Vzorec: C₃H₉O₃P
CAS číslo: 4672-38-2
EINECS: 225-121-8
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR