Tisk…

Benzylmagnesium chloride ∼16% w/w 1 mol/l diethyl ether, Sigma-Aldrich®

Dodavatel: Merck


MQ100

302759-800ML 302759-100ML
SIAL302759-800MLEA 8010 CZK
SIAL302759-800ML SIAL302759-100ML
Benzylmagnesium chloride ∼16% w/w 1 mol/l diethyl ether, Sigma-Aldrich®
Benzylmagnesium chloride
Vzorec: C₇H₇ClMg
MW: 150.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000469
CAS číslo: 6921-34-2
EINECS: 230-039-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR