Tisk…

3,6-Dichloropyridazine-4-carboxylic acid, Sigma-Aldrich®

Dodavatel: Merck


MQ100

297755-5G
SIAL297755-5GEA 3030 CZK
SIAL297755-5G
3,6-Dichloropyridazine-4-carboxylic acid, Sigma-Aldrich®
3,6-Dichloropyridazine-4-carboxylic acid
Vzorec: C₅H₂Cl₂N₂O₂
MW: 192.99
MDL Number: MFCD00011575
CAS číslo: 51149-08-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR