Tisk…

L(+)-Lysine monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: L-(+)-Lysine, Hydrate, (S)-(+)-2,6-Diaminohexanoic acid monohydrate, (S)-(+)-Lysine monohydrate, L-Lysine hydrate, (S)-(+)-2,6-Diaminocaproic acid monohydrate, H-Lys-OH monohydrate, K monohydrate, H-Lys-OH·H₂O, Lys monohydrate

M-Clarity™ quality level = MQ100

282677-25G
SIAL282677-25GEA 2100 CZK
SIAL282677-25G
L(+)-Lysine monohydrate, Sigma-Aldrich®
L(+)-Lysine monohydrate
Vzorec: H₂N(CH₂)₄CH(NH₂)CO₂H·H₂O
Bod tání: 225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151035
CAS číslo: 39665-12-8
EINECS: 200-294-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR