Tisk…

Platina, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Platinium

M-Clarity™ quality level = MQ100

267201-2G 267201-400MG
SIAL267201-2GEA 31660 CZK
SIAL267201-2G SIAL267201-400MG
Platina, Sigma-Aldrich®
Platina
Vzorec: Pt
Bod varu: 3827 °C (1013 hPa)
Bod tání: 1769 °C
Hustota: 21,447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS číslo: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR