Tisk…

Methyl phenyl sulphoxide, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

261696-25G 261696-5G
SIAL261696-25GEA 7250 CZK
SIAL261696-25G SIAL261696-5G
Methyl phenyl sulphoxide, Sigma-Aldrich®
Methyl phenyl sulphoxide
Vzorec: C₇H₈OS
MDL Number: MFCD00002088
CAS číslo: 1193-82-4
EINECS: 214-781-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR