Tisk…

Benzoyl isothiocyanate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

261653-25G 261653-5G
SIAL261653-25GEA 6770 CZK
SIAL261653-25G SIAL261653-5G
Benzoyl isothiocyanate, Sigma-Aldrich®
Benzoyl isothiocyanate
Vzorec: C₈H₅NOS
Bod varu: 252 °C (1013 hPa)
Bod tání: –1 °C
Hustota: 1,197 g/cm³ (16 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004815
CAS číslo: 532-55-8
EINECS: 208-540-0
UN: 2810
ADR: 6.1,III
Merck Index: 13,01115

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR