Tisk…

1,2-Epoxyoctan, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ100

260258-100G 260258-25G
SIAL260258-100GEA 20740 CZK
SIAL260258-100G SIAL260258-25G
1,2-Epoxyoctan, Sigma-Aldrich®
1,2-Epoxyoctan
Vzorec: C₈H₁₆O
Bod varu: 42 °C (0,2 mmHg)
Hustota: 0,8395 g/cm³ (0 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005157
CAS číslo: 2984-50-1
EINECS: 221-047-5
UN: 3271
ADR: 3,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR