Tisk…

Methyl-o-anisate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Methoxybenzoic acid methyl ester, Methyl 2-methoxybenzoate, o-Anisic acid methyl ester, 2-Anisic acid methyl ester

M-Clarity™ quality level = MQ100

259659-25G
SIAL259659-25GEA 1250 CZK
SIAL259659-25G
Methyl-o-anisate, Sigma-Aldrich®
Methyl-o-anisate
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
MDL Number: MFCD00008423
CAS číslo: 606-45-1
EINECS: 210-118-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR