Tisk…

Ethyl phenylglyoxylate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

258911-25G 258911-5G
SIAL258911-25GEA 3050 CZK
SIAL258911-25G SIAL258911-5G
Ethyl phenylglyoxylate, Sigma-Aldrich®
Ethyl phenylglyoxylate
Vzorec: C₁₀H₁₀O₃
MDL Number: MFCD00009120
CAS číslo: 1603-79-8
EINECS: 216-504-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR