Tisk…

5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

252182-500MG
SIAL252182-500MGEA 2950 CZK
SIAL252182-500MG
5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II), Sigma-Aldrich®
5,10,15,20-Tetraphenyl-21H,23H-porphine copper(II)
Vzorec: C₄₄H₂₈CuN₄
Hustota: 1,276 g/cm³ (20 °C)
MDL Number: MFCD00148862
CAS číslo: 14172-91-9
EINECS: 213-025-9

Objednat

Výsledky specifikačního testu

Appearance (Color) Faint Purple to Dark Purple
Appearance (Form) Powder or Crystals
Infrared Spectrum Conforms to Structure
Wavelength 414 - 420 nm
Extinction Coefficient ≥ 390000 (c = 0.001g/l, Toluene)
Carbon 75.8 - 80.5 %
Nitrogen 7.5 - 8.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR