Tisk…

1-Naphthoyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

250252-10G 250252-50G
SIAL250252-10GEA 1360 CZK
SIAL250252-10G SIAL250252-50G
1-Naphthoyl chloride, Sigma-Aldrich®
1-Naphthoyl chloride
Vzorec: C₁₁H₇ClO
MW: 190.63 g/mol
MDL Number: MFCD00004002
CAS číslo: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR