Tisk…

4-Chloro-2-fluorotoluene, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

247707-25G
SIAL247707-25GEA 2460 CZK
SIAL247707-25G
4-Chloro-2-fluorotoluene, Sigma-Aldrich®
4-Chloro-2-fluorotoluene
Vzorec: C₇H₆ClF
Bod varu: 153 °C (1013 hPa)
Hustota: 1,184 g/cm³ (20 °C)
MDL Number: MFCD00000571
CAS číslo: 452-75-5
EINECS: 207-210-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR